accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4963-47-7

IUPAC Name: tris(3-aminopropyl)amine
Synonyms: tris(3-aminopropyl)amine
Molecular Weight (g/mol): 188.32
Molecular Formula: C9H24N4
InChi Key: QMXSDTGNCZVWTB-UHFFFAOYSA-N
SMILES: NCCCN(CCCN)CCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results