accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49638-64-4

IUPAC Name: bis(methylsulfanyl)-2H-1,3-dithiole-2-thione
Synonyms: bis(methylsulfanyl)-1,3-dithiole-2-thione
Molecular Weight (g/mol): 226.4
Molecular Formula: C5H6S5
InChi Key: HOFVXSUZSDYZSA-UHFFFAOYSA-N
SMILES: CSC1=C(SC)SC(=S)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results