accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCO

CAS RN 49647-20-3

IUPAC Name: 1-(4-isocyanatophenyl)ethan-1-one
Synonyms: 1-(4-isocyanatophenyl)ethanone
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: MOZSVHZOUDIZMF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results