accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClN

CAS RN 4965-33-7

IUPAC Name: 7-chloro-2-methylquinoline
Synonyms: 7-chloro-2-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: WQZQFYRSYLXBGP-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC(Cl)=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results