accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrNBr

CAS RN 49669-22-9

IUPAC Name: 6,6'-dibromo-2,2'-bipyridine
Synonyms: 6,6'-dibromo-2,2'-bipyridine
Molecular Weight (g/mol): 313.98
Molecular Formula: C10H6Br2N2
InChi Key: WZVWSOXTTOJQQQ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)C1=NC(Br)=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results