accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONNNBOH3CH3CH3CCH3O

CAS RN 496786-98-2

IUPAC Name: tert-butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol): 389.3
Molecular Formula: C20H32BN3O4
InChi Key: JWUBVPJWWYYRLJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results