accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNBr

CAS RN 496807-37-5

IUPAC Name: 2-(4-bromophenyl)-6-methyl-1H-1,3-benzodiazole
Synonyms: 2-(4-bromophenyl)-5-methyl-3H-1,3-benzodiazole
Molecular Weight (g/mol): 287.16
Molecular Formula: C14H11BrN2
InChi Key: LWVJBMHQRDXWGN-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(NC2=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results