accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 497-06-3

OHHOH2C

CAS RN 497-06-3

IUPAC Name: but-3-ene-1,2-diol
Synonyms: 3-butene-1,2-diol
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: ITMIAZBRRZANGB-UHFFFAOYNA-N
SMILES: OCC(O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results