accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(R)OH2NOH2NO

CAS RN 49705-98-8

IUPAC Name: (1R,2S)-1-benzyl-1-carbamoyl-2-hydroxypropyl carbamate
Synonyms: (1R,2S)-1-benzyl-1-carbamoyl-2-hydroxypropyl carbamate
Molecular Weight (g/mol): 252.27
Molecular Formula: C12H16N2O4
InChi Key: XYJIXTMTYOGXGG-QPUJVOFHSA-N
SMILES: C[C@H](O)[C@@](CC1=CC=CC=C1)(OC(N)=O)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results