accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFCl

CAS RN 49713-56-6

IUPAC Name: 4-chloro-6-(trifluoromethyl)quinoline
Synonyms: 4-chloro-6-(trifluoromethyl)quinoline
Molecular Weight (g/mol): 231.6
Molecular Formula: C10H5ClF3N
InChi Key: FTNQANJWBFKPIP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2N=CC=C(Cl)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results