accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49715-04-0

IUPAC Name: chloro({[(chlorooxy)sulfonyl]oxy})methane
Synonyms: chloro[(chlorooxysulfonyl)oxy]methane
Molecular Weight (g/mol): 180.98
Molecular Formula: CH2Cl2O4S
InChi Key: ZFEQQZXFULOKEK-UHFFFAOYSA-N
SMILES: ClCOS(=O)(=O)OCl
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)