accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOH2NHCl

CAS RN 49755-94-4

IUPAC Name: 2-amino-N-methylacetamide hydrochloride
Synonyms: 2-amino-N-methylacetamide hydrochloride
Molecular Weight (g/mol): 124.57
Molecular Formula: C3H9ClN2O
InChi Key: ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
SMILES: Cl.CNC(=O)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)