accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49763-65-7

IUPAC Name: 4-pentylbenzoyl chloride
Synonyms: 4-pentylbenzoyl chloride
Molecular Weight (g/mol): 210.7
Molecular Formula: C12H15ClO
InChi Key: FBBRKYLXMNQFQU-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)