accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49763-66-8

IUPAC Name: 4-octylbenzaldehyde
Synonyms: 4-octylbenzaldehyde
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: IHKVZPVHTKOSLW-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results