accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHOO(E)

CAS RN 49763-96-4

IUPAC Name: (1E)-1-(2H-1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ol
Synonyms: stiripentol stiripentol (substance) estiripentol stiripentolum stiripentol (product)
Molecular Weight (g/mol): 234.3
Molecular Formula: C14H18O3
InChi Key: IBLNKMRFIPWSOY-FNORWQNLNA-N
SMILES: CC(C)(C)C(O)\C=C\C1=CC=C2OCOC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results