accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49764-63-8

IUPAC Name: 4,5-dibromobenzene-1,2-diamine
Synonyms: 4,5-dibromobenzene-1,2-diamine
Molecular Weight (g/mol): 265.94
Molecular Formula: C6H6Br2N2
InChi Key: TTXGKCVKGXHPRI-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(Br)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results