accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 49769-76-8

IUPAC Name: 1,3-dimethyl 2-cyclohexylpropanedioate
Synonyms: 1,3-dimethyl 2-cyclohexylpropanedioate
Molecular Weight (g/mol): 214.26
Molecular Formula: C11H18O4
InChi Key: CULITIDIVVTWIV-UHFFFAOYSA-N
SMILES: COC(=O)C(C1CCCCC1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results