CAS RN 49769-78-0
IUPAC Name:
1,3-dimethyl 2-benzylpropanedioate
Synonyms:
1,3-dimethyl 2-benzylpropanedioate
Molecular Weight (g/mol):
222.24
Molecular Formula:
C12H14O4
InChi Key:
ZMYJSOIMFGAQRQ-UHFFFAOYSA-N
SMILES:
COC(=O)C(CC1=CC=CC=C1)C(=O)OC
1
–
1
of
1
results
1
Dimethyl Benzylmalonate 95.0+%, TCI America™
CAS: 49769-78-0 Molecular Formula: C12H14O4 Molecular Weight (g/mol): 222.24 MDL Number: MFCD01445471 InChI Key: ZMYJSOIMFGAQRQ-UHFFFAOYSA-N Synonym: Benzylmalonic Acid Dimethyl Ester PubChem CID: 10900856 IUPAC Name: 1,3-dimethyl 2-benzylpropanedioate SMILES: COC(=O)C(CC1=CC=CC=C1)C(=O)OC