accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4979-79-7

IUPAC Name: 4-chloro-2-phenylquinoline
Synonyms: 4-chloro-2-phenylquinoline
Molecular Weight (g/mol): 239.7
Molecular Formula: C15H10ClN
InChi Key: GLVDSTVYOFXBKT-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=NC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results