CAS RN 497934-76-6
IUPAC Name:
disodium trihydrate [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms:
disodium trihydrate [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol):
763.45
Molecular Formula:
C21H33N7Na2O17P2
InChi Key:
NKGIRZCFNXSJBB-ZVYMQMITSA-L
SMILES:
O.O.O.[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
1
–
1
of
1
results
1
beta-Nicotinamide adenine dinucleotide reduced disodium salt trihydrate, 98%
CAS: 497934-76-6 Molecular Formula: C21H27N7Na2O14P2 Molecular Weight (g/mol): 709.41 MDL Number: MFCD00036200 InChI Key: QRGNQKGQENGQSE-WUEGHLCSSA-L Synonym: nadh disodium salt trihydrate,beta-nicotinamide adenine dinucleotide reduced disodium salt trihydrate PubChem CID: 101609545 IUPAC Name: disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate SMILES: [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O