accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 498-23-7

IUPAC Name: (2Z)-2-methylbut-2-enedioic acid
Synonyms: citraconic acid|mesaconate|methylfumaric acid|2-methylfumaric acid|kyselina mesakonova
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H6O4
InChi Key: HNEGQIOMVPPMNR-IHWYPQMZSA-N
SMILES: C\C(=C\C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)