accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4985-85-7

IUPAC Name: 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol): 162.23
Molecular Formula: C7H18N2O2
InChi Key: FKJVYOFPTRGCSP-UHFFFAOYSA-N
SMILES: NCCCN(CCO)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results