accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHO

CAS RN 499-05-8

IUPAC Name: 4-oxo-4H-pyran-2-carboxylic acid
Synonyms: 4-oxopyran-2-carboxylic acid
Molecular Weight (g/mol): 140.09
Molecular Formula: C6H4O4
InChi Key: UFMDRPPBUAPWBK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C=CO1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)