accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOH

CAS RN 499-44-5

IUPAC Name: 2-hydroxy-6-(propan-2-yl)cyclohepta-2,4,6-trien-1-one
Synonyms: hinokitiol 6-I-propyltropalone thujaplicin, β β-thujaplicin isopropyltropolone β-thujaplicin, sodium salt hinokitol α-thujaplicin β-thujaplicine β-isopropyltropolon
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: FUWUEFKEXZQKKA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=O)C(O)=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)