accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNOF

CAS RN 499-55-8

IUPAC Name: 3-fluorobenzohydrazide
Synonyms: 3-fluorobenzohydrazide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: UUISEXNUHLZEDB-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results