accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4991-65-5

OHSOO

CAS RN 4991-65-5

IUPAC Name: 6-hydroxy-2H-1,3-benzoxathiol-2-one
Synonyms: tioxolone tioxolona thioxalone tioxolonum tioxolon thioxolone
Molecular Weight (g/mol): 168.17
Molecular Formula: C7H4O3S
InChi Key: SLYPOVJCSQHITR-UHFFFAOYSA-N
SMILES: OC1=CC=C2SC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results