accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 499793-84-9

IUPAC Name: 1-(4-propylphenyl)-2,5-bis(thiophen-2-yl)-1H-pyrrole
Synonyms: 1-(4-propylphenyl)-2,5-bis(thiophen-2-yl)pyrrole
Molecular Weight (g/mol): 349.51
Molecular Formula: C21H19NS2
InChi Key: JFTYJFYWQQLLFG-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)N1C(=CC=C1C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results