accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3

CAS RN 4998-76-9

IUPAC Name: cyclohexanaminium chloride
Synonyms: cyclohexylammonium ion chloride
Molecular Weight (g/mol): 135.64
Molecular Formula: C6H14ClN
InChi Key: ZJUGSKJHHWASAF-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1CCCCC1
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (4)
Grade 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)