CAS RN 50-06-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Phenobarbital Solution, 1 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This certified solution standard is suitable as starting material for the preparation of calibrators and controls in phenobarbital testing by GC/MS or LC-MS/MS.