accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(R)OH(S)(R)HO(R)HO

CAS RN 50-27-1

IUPAC Name: (1R,2R,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,2,7-triol
Synonyms: estriol 16-epiestriol estratriol 雌三醇 unconjugated oestriol unconjugated estriol gynasan 3,16α,17β-estriol 16,17 epiestriol estriol product
Molecular Weight (g/mol): 288.39
Molecular Formula: C18H24O3
InChi Key: PROQIPRRNZUXQM-ZXXIGWHRSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1C[C@@H](O)[C@@H]2O
Grade 
  • (10)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (4)
  • (6)
Boiling Point 
  • (6)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (6)
  • (10)
  • (10)
  • (4)
  • (6)
  • (4)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)