accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClCl

CAS RN 50-45-3

IUPAC Name: 2,3-dichlorobenzoic acid
Synonyms: benzoic acid, 2,3-dichloro-
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: QAOJBHRZQQDFHA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)