accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)CH3(S)HNO(S)HONHO(R)NH2SS(R)HNO(S)NH2OHNO(S)NH2ONHOON(S)ONH(S)CH3H3COHNNH2O

CAS RN 50-56-6

IUPAC Name: (2S)-2-{[(2S)-1-[(4R,7S,10S,13S,16S,19R)-19-amino-13-[(2S)-butan-2-yl]-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl]formamido}-N-(carbamoylmethyl)-4-methylpentanamide
Synonyms: oxytocin ocytocin oxitocina endopituitrina oxytocine syntocinon OXT piton S pitocin (parke-davis) orasthin
Molecular Weight (g/mol): 1007.19
Molecular Formula: C43H66N12O12S2
InChi Key: XNOPRXBHLZRZKH-DSZYJQQASA-N
SMILES: CC[C@H](C)[C@@H]1NC(=O)[C@H](CC2=CC=C(O)C=C2)NC(=O)[C@@H](N)CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCC(N)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)