accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClCl

CAS RN 50-79-3

IUPAC Name: 2,5-dichlorobenzoic acid
Synonyms: 2,5-dichlorobenzoic acid
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: QVTQYSFCFOGITD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)