accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClCl

CAS RN 50-84-0

IUPAC Name: 2,4-dichlorobenzoic acid
Synonyms: 2,4-dichlorobenzoic acid C7H4cl2O2
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: ATCRIUVQKHMXSH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (6)
  • (5)
  • (6)
  • (2)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)