accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOHOH(E/Z)

CAS RN 500-65-2

IUPAC Name: 5-[2-(3-hydroxy-4-methoxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: rhapontigenin
Molecular Weight (g/mol): 258.27
Molecular Formula: C15H14O4
InChi Key: PHMHDRYYFAYWEG-UHFFFAOYSA-N
SMILES: COC1=C(O)C=C(C=CC2=CC(O)=CC(O)=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results