accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHO

CAS RN 500-76-5

IUPAC Name: 4-(4-hydroxyphenoxy)benzoic acid
Synonyms: 4-(4-hydroxyphenoxy)benzoic acid
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H10O4
InChi Key: KLXPCYHWTLAVLN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results