accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 500013-38-7

IUPAC Name: 3-({4-[(tert-butoxy)carbonyl]piperazin-1-yl}methyl)benzoic acid
Synonyms: 3-{[4-(tert-butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
Molecular Weight (g/mol): 320.39
Molecular Formula: C17H24N2O4
InChi Key: CXJBKOKDFABTQO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=CC(=C2)C(O)=O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results