accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 5002-26-6

IUPAC Name: 4-bromo-2-methyl-1,1'-biphenyl
Synonyms: 4-bromo-2-methyl-1,1'-biphenyl
Molecular Weight (g/mol): 247.14
Molecular Formula: C13H11Br
InChi Key: ZBNARPVMXYNXQQ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results