accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5002-93-7

IUPAC Name: 1-amino-3-(4-methoxyphenoxy)propan-2-ol
Synonyms: 1-amino-3-(4-methoxyphenoxy)propan-2-ol
Molecular Weight (g/mol): 197.23
Molecular Formula: C10H15NO3
InChi Key: KFQPRNVTVMCYEH-UHFFFAOYNA-N
SMILES: COC1=CC=C(OCC(O)CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results