accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrNOO

CAS RN 500298-30-6

IUPAC Name: 1-bromo-3-methoxy-2-nitrobenzene
Synonyms: 1-bromo-3-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: ORSPVBUIPTYLLO-UHFFFAOYSA-N
SMILES: COC1=C(C(Br)=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results