accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOOOCH3O

CAS RN 5003-48-5

IUPAC Name: 4-acetamidophenyl 2-(acetyloxy)benzoate
Synonyms: benorilate
Molecular Weight (g/mol): 313.31
Molecular Formula: C17H15NO5
InChi Key: FEJKLNWAOXSSNR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results