accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5006-44-0

IUPAC Name: 6-methyl-4-oxo-4H-chromene-2-carboxylate
Synonyms: 6-methyl-4-oxochromene-2-carboxylate
Molecular Weight (g/mol): 203.17
Molecular Formula: C11H7O4
InChi Key: RKIDLZFIIVDZNX-UHFFFAOYSA-M
SMILES: CC1=CC=C2OC(=CC(=O)C2=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)