accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 501-24-6

IUPAC Name: 3-pentadecylphenol
Synonyms: phenol, 3-pentadecyl-
Molecular Weight (g/mol): 304.52
Molecular Formula: C21H36O
InChi Key: PTFIPECGHSYQNR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC1=CC=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results