accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrF

CAS RN 501420-63-9

IUPAC Name: 2-(3-bromo-4-fluorophenyl)acetonitrile
Synonyms: 2-(3-bromo-4-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: ORKCKZRBHXMWBO-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results