accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5015-38-3

IUPAC Name: barium(2+) 3-(phosphonatooxy)propanenitrile
Synonyms: barium(2+) 3-(phosphonatooxy)propanenitrile
Molecular Weight (g/mol): 286.37
Molecular Formula: C3H4BaNO4P
InChi Key: MRQIDZJGQMWVQR-UHFFFAOYSA-L
SMILES: [Ba++].[O-]P([O-])(=O)OCCC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results