accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 501944-63-4

IUPAC Name: 5-(4-ethynylphenyl)-1,3-oxazole
Synonyms: 5-(4-ethynylphenyl)-1,3-oxazole
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: CYRYZDZQFDRTHD-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results