CAS RN 502-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ricca Chemical Company Farnesene, Natural Isolate, Ricca Chemical
Farnesene, Natural Isolate is a natural isolate food additive.