CAS RN 5024-21-5
IUPAC Name:
1,14-dimethyl tetradecanedioate
Synonyms:
1,14-dimethyl tetradecanedioate
Molecular Weight (g/mol):
286.41
Molecular Formula:
C16H30O4
InChi Key:
ZDJFDFNNEAPGOP-UHFFFAOYSA-N
SMILES:
COC(=O)CCCCCCCCCCCCC(=O)OC
1
–
1
of
1
results
1
Dimethyl Tetradecanedioate 96.0+%, TCI America™
CAS: 5024-21-5 Molecular Formula: C16H30O4 Molecular Weight (g/mol): 286.41 MDL Number: MFCD00673459 InChI Key: ZDJFDFNNEAPGOP-UHFFFAOYSA-N PubChem CID: 78727 IUPAC Name: 1,14-dimethyl tetradecanedioate SMILES: COC(=O)CCCCCCCCCCCCC(=O)OC