accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNONHNClClOCH3

CAS RN 502486-89-7

IUPAC Name: 1-(2,4-dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide
Synonyms: 1-(2,4-dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-(piperidin-1-yl)pyrazole-3-carboxamide
Molecular Weight (g/mol): 459.37
Molecular Formula: C23H24Cl2N4O2
InChi Key: KWDBQJRWPWTGPF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(C)C(=NN1C1=C(Cl)C=C(Cl)C=C1)C(=O)NN1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results