accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOHOH

CAS RN 5026-65-3

IUPAC Name: hexadecyl 3,4,5-trihydroxybenzoate
Synonyms: hexadecyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol): 394.55
Molecular Formula: C23H38O5
InChi Key: TYCUSKFOGZNIBO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results